Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Listeria Enrichment Broth

Catalog No. OXCM0862B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0862B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0862B Supplier Thermo Scientific™ Supplier No. CM0862B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Oxoid Listeria Enrichment Broth is used for the enrichment of Listeria species.

Oxoid Listeria Enrichment Broth is used for the enrichment of Listeria species.

Also available Listeria Selective Supplement, Part No. SR0140E or SR0141E and Modified Listeria Selective Enrichment Supplement, Part No. SR0206E or SR0206B.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: FDA, IDF.
Description Listeria Enrichment Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 13.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.