Learn More
Thermo Scientific™ Oxoid™ ONE Broth-Listeria
Description
Oxoid ONE Broth-Listeria is a selective enrichment broth for Listeria species in food samples in 24 hours. Novel enrichment broth balances a mix of peptones, carbohydrates and salts for optimal resuscitation, recovery and growth of even low numbers of Listeria spp.
Requires ONE Broth-Listeria Supplement, Part No. SR00234E.
ONE Broth - Listeria: Typical Formulation
- Peptone: 26g/L; Carbohydrate mix: 6g/L; Salt mix: 10g/L with a final pH of 7.4 ±0.2 at 25°C
Appearance:
- Dehydrated medium: straw coloured, free-flowing powder
- Selective supplement: orange/yellow, freeze-dried pellet
- Prepared medium: dark straw coloured, clear liquid
Formulation Directions
- Suspend 22g of ONE Broth-Listeria Base in 500ml (44g/L) of distilled water
- Mix well and sterilize by autoclaving at 121°C for 15 minutes
- Cool the medium to below 50°C and aseptically add the contents of one vial of ONE Broth-Listeria Selective Supplement (Mfr. No. SR0234) reconstituted as directed
Testing Protocol
- Add 25g of food sample to 225mL of ONE Broth-Listeria and stomach for a minimum of 30 seconds to mix the sample
- Incubate the broth without agitation at 30°C for 24 hours
- Gently agitate the bag then, using a microbiological loop, remove 10μL and inoculate onto a suitable agar medium e.g. Brilliance Listeria Agar (Mfr. No. CM1080, SR0222, and SR0228) and incubate appropriately (37°C for 24-48 hours)
- Confirm presumptive colonies on the agar plates as Listeria monocytogenes or Listeria spp. by appropriate methods e.g. Gram-stain, catalase, oxidase, Oxoid Listeria Latex Test Kit (Mfr. No. DR1126)
- Oxoid O.B.I.S. mono (Mfr. No. ID0600), Microbact Listeria 12L (Mfr. No. MB1128)
Specifications
Specifications
| Description | ONE Broth-Listeria |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 2.5 kg |
| Yield | For 57L medium |
| Species | Listeria |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.