Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Oxoid™ Proteose Peptone
Click to view available options
Quantity:
500 g
Description
Thermo Scientific™ Oxoid Proteose Peptone is a specialty peptone containing proteoses as defined in the USP.
Particularly suitable for use in toxin production and fermentation applications or in culture media for bacteria with wide-ranging nutritional requirements.
- Particularly suitable for use in toxin production and in culture media for bacteria with wide-ranging nutritional requirements
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: USP. |
| Content And Storage | Dehydrated Powder, Ambient & Dry Storage |
| Form | Dehydrated Powder |
| Product Type | Peptone |
| For Use With (Application) | Culture media or fermentation processes |
| Quantity | 500 g |
| Protein Source | Pork / Beef / Soy |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction