Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ VCAT Selective Supplement

Catalog No. p-4522620
Click to view available options
Quantity:
500 mL of Base
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for the preparation of New York City Medium for the isolation of Neisseria gonorrhoeae.

Thermo Scientific™ Oxoid VCAT Selective Supplement is used for the preparation of New York City Medium for the isolation of Neisseria gonorrhoeae.

Add to GC Agar Base, Part No. CM0367B
TRUSTED_SUSTAINABILITY

Specifications

Description VCAT, 10 Vials, each supplements 500mL of base
Form Liquid
Product Type Selective Supplement
Quantity 500 mL of Base
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.