Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxone(R), Monopersulfate Compound, Spectrum™ Chemical
SDP

Catalog No. 18604921 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-921 5 kg Poly Pail
18-604-919 100 g Poly Bottle
18-604-920 1 kg Poly Bottle
18-604-922 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-921 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. O13065KG
Add to Cart
Add to Cart

O1306, 37222-66-5, 2KHSO5·KHSO4·K2SO4

Spectrum™ Chemical Oxone(R), Monopersulfate Compound , also known as potassium peroxymonosulfate or MPS, is the potassium salt of peroxymonosulfuric acid and used as an oxidizing agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 37222-66-5
Molecular Formula H2K4O13S3
Molecular Weight (g/mol) 462.58
InChI Key TZTINBNQGPHMBC-UHFFFAOYSA-J
IUPAC Name tetrapotassium oxidanesulfonoperoxoate hydrogen sulfate sulfate
SMILES [K+].[K+].[K+].[K+].OS([O-])(=O)=O.[O-]S([O-])(=O)=O.OS(=O)(=O)O[O-]

Specifications

CAS Max % 1
Quantity 5 kg
UN Number UN3260
Grade Reagent
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.