Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXPAT Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NB11060511

 View more versions of this product

Catalog No. NB11060511


Only null left
Add to Cart

Description

Description

OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

OXPAT
Polyclonal
Unconjugated
PBS and 30% Glycerol with 0.1% Sodium Azide
PLIN5
A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProt# Q8BVZ1]
Affinity Purified
RUO
Primary
Human and mouse. Immunogen sequence has 89% homology to rat and 75% homology to bovine.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot
1.01 mg/mL
Western Blot 0.5-2 ug/ml, Simple Western 10 ug/ml
lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5
Rabbit
50 kDa
0.1 mL
Diabetes Research, Lipid and Metabolism, Lipid Droplets
440503
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only