Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXPAT Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060511H
Description
OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
OXPAT | |
Polyclonal | |
Western Blot, Simple Western | |
lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5 | |
Rabbit | |
50 kDa | |
0.1 mL | |
Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
440503 | |
Human, Mouse | |
IgG |
Western Blot | |
HRP | |
PBS with No Preservative | |
PLIN5 | |
A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProt# Q8BVZ1] | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Immunogen sequence has 89% homology to rat and 75% homology to bovine. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction