Learn More
Invitrogen™ OXPAT Polyclonal Antibody

Description
The target sequence has 94% sequence homology with sheep and porcine and 84% sequence homology with rat. Suggested positive control: human and mouse heart lysate.
Specifications
Specifications
| Antigen | OXPAT |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Plin5 |
| Gene Accession No. | A6QLL0, Q00G26, Q8BVZ1 |
| Gene Alias | 2310076L09Rik; AI415325; AW109675; lipid droplet associated protein; lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDA5; LSDP5; Mldp; Myocardial LD protein; Oxpat; Pat1; PAT-1; perilipin 5; perilipin-5; Plin5 |
| Gene Symbols | Plin5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.