Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OxPhos Human WB Antibody Cocktail
GREENER_CHOICE

Catalog No. 458199
Encompass_Preferred
Change view
Click to view available options
Quantity:
360 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-819-9 360 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-819-9 Supplier Invitrogen™ Supplier No. 458199
Add to Cart
Edge
Add to Cart

Antibody Cocktail

Contains 5 monoclonal antibodies, one each against Complex I subunit [NDUFB8 (20E9DH10C12)], Complex II subunit 30kDa [SDHB (21A11AE7)], Complex III subunit Core 2 [UQCRC2 (13G12AF12BB11)], Complex IV subunit II [MTCO2 (12C4F12)], and ATP synthase subunit alpha [ATP5A (15H4C4)] as an optimized premixed cocktail. The cocktail is suitable for Western Blotting analysis of the relative levels of the 5 OXPHOS complexes in mitochondrial preparations from human sources.

Altered levels of assembly can arise from mutations in individual subunits, mutations in assembly factors for the complex(es), mtDNA depletion or as a result of physiological and or pathological changes e.g. hormone treatment, exercise, diet or oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OxPhos Human WB
Applications Western Blot
Classification Cocktail
Clone Cocktail
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene Alias 12C4F12; 13G12AF12BB11; 15H4C4; 20E9DH10C12; 21A11AE7; OxPhos Complex
Host Species Mouse
Purification Method IgG fraction
Quantity 360 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.