Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | OXR1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
OXR1 Polyclonal specifically detects OXR1 in Human samples. It is validated for Western Blot.Specifications
| OXR1 | |
| Polyclonal | |
| Rabbit | |
| Q8N573 | |
| 55074 | |
| Synthetic peptides corresponding to OXR1(oxidation resistance 1) The peptide sequence was selected from the N terminal of OXR1. Peptide sequence VESSPSLSPVSPLSPTSSEAEFDKTTNPDVHPTEATPSSTFTGIRPARVV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ10125, FLJ38829, FLJ40849, FLJ41673, FLJ42450, FLJ45656, oxidation resistance 1, oxidation resistance protein 1, putative protein product of Nbla00307 | |
| OXR1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title