Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582970 Supplier Invitrogen™ Supplier No. PA582970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDSFLHENSL HQEESQKENM PCGETAEFKQ KQSVNKGKQG KEQNQDSQTE AEELRKLWKT HTMQQTKQQR ENIQQVS Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 58%.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OXR1
Gene Accession No. Q8N573
Gene Alias 2210416C20Rik; C7; C7B; Gm1238; Nbla00307; nucleolar protein C7; oxidation resistance 1; oxidation resistance protein 1; Oxr1; Protein C7; putative protein product of Nbla00307; TBC/LysM-associated domain containing 3; TLDC3
Gene Symbols OXR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OXR1. Recombinant protein control fragment (Product #RP-93874).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.