Learn More
OXR1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302035AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May be involved in protection from oxidative damage. Ohira M., Cancer Lett. 197:63-68(2003). The German cDNA consortium; Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases.Specifications
OXR1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8N573 | |
OXR1 | |
Between 175 and 225 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OXR1 | |
oxidation resistance protein 1, putative protein product of Nbla00307 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55074 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.