Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXSM Monoclonal Antibody (OTI4E10), TrueMAB™, OriGene
SDP

Catalog No. 501679136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9136 Supplier OriGene Supplier No. TA802185
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a beta-ketoacyl synthetase. The encoded enzyme is required for elongation of fatty acid chains in the mitochondria. Alternatively spliced transcript variants have been described.

Specifications

Antigen OXSM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OXSM
Gene Accession No. Q9NWU1
Gene Alias CEM1, FASN2D, KASI, KS
Gene Symbols OXSM
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 78-343 of human OXSM produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54995
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.