Learn More
OXSM Mouse anti-Human, Clone: OTI3A4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802203

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a beta-ketoacyl synthetase. The encoded enzyme is required for elongation of fatty acid chains in the mitochondria. Alternatively spliced transcript variants have been described.Specifications
OXSM | |
Monoclonal | |
Unconjugated | |
OXSM | |
CEM1, FASN2D, KASI, KS | |
Mouse | |
Affinity Chromatography | |
RUO | |
54995 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3A4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NWU1 | |
OXSM | |
Human recombit protein fragment corresponding to amino acids 78-343 of human OXSM produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.