Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXSM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582755 Supplier Invitrogen™ Supplier No. PA582755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQSEADQVAT GVAIGMGMIP LEVVSETALN FQTKGYNKVS PFFVPKILVN MAAGQVSIRY KLKGPNHAVS TACTTGAHAV GDSFRFIAHG DADVMVAGGT DSCISPLSLA GF Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 93%.

This gene encodes a beta-ketoacyl synthetase. The encoded enzyme is required for elongation of fatty acid chains in the mitochondria. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXSM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OXSM
Gene Accession No. Q9D404, Q9NWU1
Gene Alias 3-ketoacyl-acyl carrier protein synthase; 3-oxoacyl-[acyl-carrier-protein] synthase, mitochondrial; 3-oxoacyl-ACP synthase, mitochondrial; 3-oxoacyl-acyl-carrier-protein synthase; 4933425A18Rik; acyl-malonyl acyl carrier protein-condensing enzyme; beta-ketoacyl synthase; beta-ketoacyl-ACP synthase; C80494; FASN2D; KASI; KS; Oxsm; RGD1311092; type II mitochondrial beta-ketoacyl synthase
Gene Symbols OXSM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OXSM. Recombinant protein control fragment (Product #RP-93153).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289934, 54995, 71147
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.