Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXSR1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077321AF350
Description
OXSR1 Polyclonal antibody specifically detects OXSR1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
OXSR1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.1, KIAA1101OSR1serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, oxidative-stress responsive 1 | |
The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human oxidative-stress responsive 1 using the numbering given in entry NP_005100.1 (GeneID 9943). | |
0.1 mL | |
Protein Kinase | |
9943 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction