Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXSR1 Antibody (OTI1F3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273177F
Description
OXSR1 Monoclonal specifically detects OXSR1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry.Specifications
OXSR1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
OXSR1 | |
Full-length protein expressed in 293T cell transfected with human OXSR1 expression vector. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Immunohistochemistry | |
OTI1F3 | |
Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.1, KIAA1101OSR1serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, oxidative-stress responsive 1 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
9943.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction