Learn More
OXSR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 156111AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0349
Only null left
Add to Cart
Description
Description
Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.
Specifications
Specifications
OXSR1
Polyclonal
Unconjugated
OXSR1
KIAA1101, OSR1, oxidative stress responsive 1, OXSR1
Rabbit
Antigen Affinity Chromatography
RUO
108737, 316064, 9943
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95747, Q6P9R2
Oxsr1
OXSR1 Fusion Protein Ag8002
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.