Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXTR Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP276347
Description
OXTR Polyclonal specifically detects OXTR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
OXTR | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
OXTR | |
Peptide with sequence C-LRTTRQKHSRL corresponding to internal region according to NP_000907.2. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Flow Cytometry, Peptide ELISA Detection limit 1:64000 | |
OT-R, oxytocin receptor | |
Goat | |
Affinity Purified | |
Cancer, GPCR, Neuroscience, Neurotransmission | |
5021.0 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction