Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXTR Antibody (SR1739), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32211450UL
Description
OXTR Monoclonal antibody specifically detects OXTR in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
OXTR | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1739 | |
Western Blot 1:500-1:2000 | |
OT-R, oxytocin receptor | |
A synthesized peptide derived from human OXTR (Uniprot #: P30559) | |
50 μg | |
Cancer, GPCR, Neuroscience, Neurotransmission | |
5021 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction