Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beckman Coulter™ Oxycodone Calibrator
Shop All Beckman Coulter Products
Click to view available options
Concentration:
1,000 ng/mL
100 ng/mL
300 ng/mL
500 ng/mL
Description
- For use with Beckman Coulter analyzers

Specifications
Specifications
Concentration | 100 ng/mL |
For Use With (Equipment) | Beckman Coulter Analyzers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction