Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Oxycodone Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175335
Description
Working Concentration: 1/8K. Sensitivity: 10 ng/mL Oxycodone produces 39% inhibition in a competitive ELISA employing Oxycodone (N) polyclonal antibody.
Oxycodone is a semi-synthetic, morphine-like opioid alkaloid with analgesic activity. Oxycodone exerts its analgesic activity by binding to the mu-receptors in the central nervous system (CNS), thereby mimicking the effects of endogenous opioids. Binding of the opiate receptor stimulates the exchange of GTP for GDP on the G-protein complex and inhibits adenylate cyclase, thereby preventing cAMP production. Subsequently, the release of nociceptive neurotransmitters, such as substance P, gamma-aminobutyric acid (GABA), dopamine, acetylcholine, and noradrenaline, is inhibited. Oxycodone also inhibits the release of vasopressin, somatostatin, insulin, and glucagon. In addition, oxycodone closes N-type voltage-gated calcium channels and opens G-protein-coupled inwardly rectifying potassium channels resulting in hyperpolarization and reduction of neuronal excitability. [National Center for Biotechnology Information. PubChem Compound Database; CID=5284603].
Specifications
Oxycodone | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.2 | |
Sheep | |
Purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
C18H21NO4; Dihydrohydroxycodeinone; Dihydrone; Dihydroxycodeinone; Oxycodeinone; Oxycontin | |
Oxycodone(N)-BSA. | |
1 mg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction