Learn More
Oxygen-regulated protein 1 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | Oxygen-regulated protein 1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | RP1 |
| Gene Accession No. | P56715, P56716 |
| Gene Alias | Dcdc3; DCDC4A; mG145; ORP1; Oxygen-regulated protein 1; retinitis pigmentosa 1 (autosomal dominant); retinitis pigmentosa 1 (human); retinitis pigmentosa 1 homolog; Retinitis pigmentosa 1 protein; Retinitis pigmentosa RP1 protein; Retinitis pigmentosa RP1 protein homolog; Rp1; Rp1h |
| Gene Symbols | RP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.