Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxyntomodulin Antibody (46), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261947AF350
Description
Oxyntomodulin Monoclonal specifically detects Oxyntomodulin in Human samples. It is validated for ELISA.Specifications
Oxyntomodulin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Thsi antibody is specific for C-terminus of human oxyntomodulin. Does not cross-react with glucagon | |
Store at 4°C in the dark. |
ELISA | |
46 | |
ELISA | |
GCG, GLP1, GLP2, GRPP | |
Synthetic oxyntomodulin 31-37 coupled to carrier | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction