Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OXYR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144410
Description
OXYR Polyclonal Antibody for Western Blot, ELISA
Hydrogen peroxide sensor. Activates the expression of a regulon of hydrogen peroxide-inducible genes such as katG, gor, ahpC, ahpF, oxyS (a regulatory RNA), dps, fur and grxA. OxyR expression is negatively autoregulated by binding to a 43 bp region upstream of its own coding sequence. OxyR is inactivated by reduction of its essential disulfide bond by the product of GrxA, itself positively regulated by OxyR. Has also a positive regulatory effect on the production of surface proteins that control the colony morphology and auto-aggregation ability.
Specifications
OXYR | |
Polyclonal | |
Unconjugated | |
oxyR | |
momR; mor | |
Rabbit | |
Protein G | |
RUO | |
75203207 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.04 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P0ACQ4 | |
oxyR | |
Recombinant Escherichia coli (strain K12) OXYR protein (65-305aa). | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction