Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytetracycline, 96%

Catalog No. p-7042405
Click to view available options
Quantity:
10 g
50 g
100 g

Oxytetracycline, CAS # 79-57-2, shows antibacterial, anti-inflammatory, and anti-proliferative activities. It can also inhibit mitochondrial protein synthesis. | CAS: 79-57-2 | C22H26N2Na2O11 | 540.43 g/mol

Oxytetracycline is an anti-inflammatory, antibiotic, and anti-proliferative agent shown to inhibit mitochondrial protein synthesis. Mechanistic studies show that this compound reduces cytochrome c activity which eventually causes cell growth arrest. Additionally, Oxytetracycline has demonstrated the ability to reduce murine bronchial inflammation. The anti-inflammatory characteristics of this agent are due to its ability to cause a decrease in IL-4, IL-5, IL-13, immune cells, and the ovalbumin-specific IgE. Data suggests that Oxytetracycline is also an inhibitor of NF-κB activation via phosphorylation and degradation of IκB-α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Oxytetracycline is a compound with broad-spectrum antibacterial activity. Furthermore, it also shows anti-inflammatory and anti-proliferative activities
  • It can inhibit mitochondrial protein synthesis in both gram-positive and gram-negative bacteria

Application

  • Oxytetracycline is active against several species including Mycoplasma pneumonia, Pasteurella pestis, Escherichia coli, Haemophilus influenza, and Diplococcus pneumonia
  • In laboratory experiments, oxytetracycline can be used to evaluate the acquisition of oxytetracycline-resistance genes, which are associated with the development of antibiotic resistance
  • It can reduce cytochrome c activity, causing cell growth arrest
  • It has been shown to reduce murine bronchial inflammation
  • Oxytetracycline exerts its anti-inflammatory activity by decreasing the levels of IL-4, IL-5, IL-13 immune cells, and the Ovalbumin-specific IgE
  • Oxytetracycline can be used to inhibit NF-κB activation via phosphorylation and degradation of IκB-α
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-57-2
Molecular Formula C22H26N2Na2O11
Molecular Weight (g/mol) 540.43
MDL Number MFCD00003700
InChI Key OLRBUPVCLOPDOT-GETZKYKJSA-M
PubChem CID 54675779
IUPAC Name disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
SMILES O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O

Specifications

Type Oxytetracycline
Physical Form Powder
Melting Point 184°C to 185°C
Color Tan to Yellow
Quantity 10 g
Merck Index 14,6976
Solubility Information Sparingly soluble in aqueous solution at 0.6 mg/mL
Formula Weight 460.43
Percent Purity ≥98%
Chemical Name or Material Oxytetracycline
Hazard Category H302-H312-H319-H332-H360
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P501c
EINECSNumber 201-212-8
RTECSNumber QI7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.