Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytetracycline HCl, G-Biosciences
SCI_ED

Catalog No. S157216
Click to view available options
Quantity:
10 g
50 g

Oxytetracycline is a tetracycline antibiotic derived from Streptomyces rimosus. It targets gram-positive and gram-negative bacteria and several Mycoplasma species.

Oxytetracycline has been used to study its resistance gene, otrA and in fluorescence analysis of liver cells. Additionally, oxytetracycline is used as a selective agent in OGYE, a selective medium for yeasts and molds. It has been used to prevent fire blight in apples and pears.

Chemical Identifiers

Grade Molecular Biology

Specifications

CAS 2058-46-0
Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISP 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Molecular Weight (g/mol) 496.89 g/mol
Quantity 50 g
Percent Purity >97%
Chemical Name or Material Oxytetracycline HCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.