Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxytocin/Neurophysin I Antibody (OTI5G4), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273178R
Description
Oxytocin/Neurophysin I Monoclonal specifically detects Oxytocin/Neurophysin I in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Oxytocin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
OXT | |
Recombinant protein fragment corresponding to amino acids 20-125 of human Oxytocin (NP_000906) produced in E.coli. | |
0.1 mL | |
5020.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI5G4 | |
Western Blot | |
MGC126890, MGC126892, neurophysin I, OT, OT-NPI, oxytocin, prepro- (neurophysin I), oxytocin, prepropeptide, oxytocin-neurophysin 1, oxytocin-neurophysin I, preproprotein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction