Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytocin R Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982654
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2654 Supplier Bioss Supplier No. BS1314R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The oxytocin receptor (OXTR) belongs to the G-protein coupled receptor family. Its activity is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. The receptor is made up of 389 amino acids and has a molecular weight of ˜43,000 kDa. The oxytocin-oxytocin receptor system plays an important role as an inducer of uterine contractions during parturition and of milk ejection. Oxytocin receptors are expressed by the myoepithelial cells of the mammary gland, and in both the myometrium and endometrium or the uterus at the end of pregnancy. Additionally, oxytocin receptors are present in the central nervous system, mediating a variety of behaviors, including maternal behavior, bonding, social memory and recognition, sexual and aggressive behaviors, and stress and anxiety.

Specifications

Antigen Oxytocin R
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Oxtr
Gene Accession No. P30559
Gene Alias Otr; OT-R; OTR1; oxcytocin receptor; Oxtr; Oxt-r; Oxytocin receptor
Gene Symbols Oxtr
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse oxytocin receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5021
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.