Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Oxytocin Receptor, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1735MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1735MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1735MI Supplier MilliporeSigma™ Supplier No. ABN1735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Oxytocin receptor (UniProt P97926; also known as OT-R) is encoded by the Oxtr (also known as Otr) gene (Gene ID 18430) in murine species. Oxytocin receptor (OT-R) is widely expressed in the reproductive tract, brain, kidneys, and many other organs, where it mediates the biological functions of oxytocin (OT) in mammals. Best known for their role in lactation and parturition, OT-R and OT meidate a wide variety of physiological functions, including estrous cycle, bone formation, social behaviors, as well as regulation of energy homeostasis, including food intake and thermoregulation. Central administration of OT decreases food consumption in rats, and nesfatin (a satiety hormone) increases OT secretion. Oxt-null and Oxtr-null mice display aberrant thermoregulatory function. In addition, Oxtr-knockout mice have a morphological defect in brown adipose tissue (BAT) cells and develop obesity. Human and murine oxytocin receptors are 7-transmembrane (a.a. 39-63, 75-97, 114-135, 155-175, 203-225, 275-293, 309-331) GPCRs with their N-terminal end at the extracellular side (a.a. 1-38) and C-terminal end at the cytoplasmic side (human a.a. 332-389 and mouse a.a. 332-388).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oxytocin Receptor
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene Oxtr, Otr
Gene Accession No. P97926
Host Species Rabbit
Immunogen BSA-conjugated linear peptide corresponding to a sequence from mouse oxytocin receptor N-terminal extracellular domain.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001074616
Test Specificity This polyclonal antibody specifically stained wild-type, but not Oxtr-knockout, mouse brain tissue sections (Courtesy of Dr. Moses V. Chao, Langone Medical Center, New York University, U.S.A).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.