Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxytocinase Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | Oxytocinase |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7533
|
Bethyl Laboratories, Inc
A304363AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
LNPEP, also known as insulin-responsive aminopeptidase (IRAP), was initially identified as an abundant protein that is associated with specialized endosomes termed GLUT4 storage vesicles (GSVs) in adipocytes. The luminal domain of LNPEP contains an aminopeptidase activity that is involved in peptide hormone processing and antigenic peptide processing. LNPEP is also a key regulator of GLUT4 trafficking by controlling the sorting of GLUT4 from endosomes to the GSVs.Specifications
| Oxytocinase | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| CAP, IRAP, P-LAP, PLAP, AT (4) receptor, angiotensin IV receptor, cystinyl aminopeptidase, insulin-regulated aminopeptidase, insulin-regulated membrane aminopeptidase, insulin-responsive aminopeptidase, leucyl-cystinyl aminopeptidase, otase, oxytocinase, placental leucine aminopeptidase, | |
| LNPEP | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UIQ6 | |
| 4012 | |
| Between 1 and 50 | |
| Primary | |
| 4° C | |
| LNPEP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title