Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p-Anisidine, 99%
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 56.0°C to 59.0°C |
| Color | Brown to Gray |
| Density | 1.0600g/mL |
| Boiling Point | 240.0°C to 243.0°C |
| Flash Point | 122°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | CH3OC6H4NH2 |
| Quantity | 1 kg |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause damage to organs through prolonged or repeated exposure. May cause cancer. Fatal if inhaled. Fatal in contact with skin. Fatal if swallowed. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Rinse mouth. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. |
| EINECSNumber | 203-254-2 |
| RTECSNumber | BZ5450000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction