Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Benzoquinone, 98+%

Catalog No. aaa1316222
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g

CAS: 106-51-4 | C6H4O2 | 108.096 g/mol

p-Benzoquinone is used as a dienophile in Diels-Alder cycloadditions to prepare naphthoquinones and 1,4-phenanthrenediones. It acts as a dehydrogenation reagent and an oxidizer in synthetic organic chemistry. In the Thiele-Winter reaction, it is involved in the preparation of triacetate of hydroxyquinol by reacting with acetic anhydride and sulfuric acid. It is also used in the synthesis of bromadol and to suppress double- bond migration during olefin metathesis reactions. It is used as a precursor to hydroquinone which finds application in photography and as a reducing agent and an antioxidant in rubber production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Benzoquinone is used as a dienophile in Diels-Alder cycloadditions to prepare naphthoquinones and 1,4-phenanthrenediones. It acts as a dehydrogenation reagent and an oxidizer in synthetic organic chemistry. In the Thiele-Winter reaction, it is involved in the preparation of triacetate of hydroxyquinol by reacting with acetic anhydride and sulfuric acid. It is also used in the synthesis of bromadol and to suppress double- bond migration during olefin metathesis reactions. It is used as a precursor to hydroquinone which finds application in photography and as a reducing agent and an antioxidant in rubber production.

Solubility
Soluble in water, ethanol, ether, methanol, benzene, acetone and ethyl acetate.

Notes
Light sensitive. Store in cool place. Incompatible and reacts violently with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-51-4
Molecular Formula C6H4O2
Molecular Weight (g/mol) 108.096
MDL Number MFCD00001591
InChI Key AZQWKYJCGOJGHM-UHFFFAOYSA-N
Synonym p-benzoquinone, benzoquinone, quinone, 1,4-benzoquinone, p-quinone, chinone, 2,5-cyclohexadiene-1,4-dione, cyclohexadienedione, para-quinone, 1,4-benzoquine
PubChem CID 4650
ChEBI CHEBI:16509
IUPAC Name cyclohexa-2,5-diene-1,4-dione
SMILES C1=CC(=O)C=CC1=O

Specifications

Melting Point 112°C to 115°C
Density 1.318
Boiling Point ∼180°C (sublimation)
Flash Point 77°C (171°F)
Odor Pungent
Assay Percent Range ≥98%
Quantity 100 g
UN Number UN2587
Beilstein 773967
Sensitivity Light sensitive
Merck Index 14,8074
Solubility Information Soluble in water,ethanol,ether,methanol,benzene,acetone and ethyl acetate.
Formula Weight 108.1
Percent Purity ≥98%
Chemical Name or Material p-Benzoquinone
Show More Show Less
Hazard Category H301+H331-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H331-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZOQUINONE
EINECSNumber 203-405-2
RTECSNumber DK2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.