Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Cadherin Antibody (MM0508-9V11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
| Antigen | P-Cadherin |
|---|---|
| Clone | MM0508-9V11 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Frozen), Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| P-Cadherin | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin | |
| CDH3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0508-9V11 | |
| Immunohistochemistry (Frozen), Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| CAA45177 | |
| 1001 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title