Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Glycoprotein Polyclonal Antibody, Invitrogen™

Catalog No. PIPA590029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590029 Supplier Thermo Scientific Supplier No. PA590029

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: STVRNADVIA GFDDGVIVEK GNHDELMKEK GIYFKLVTMQ TAGNEVELEN AADESKSEID ALEMSSNDSR SSLIRKRSTR RSVRGSQAQD RKLSTKEALD

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Glycoprotein
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 1.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene ABCB1
Gene Accession No. P06795, P08183, P21447
Gene Alias ABC superfamily, P-glycoprotein, ABC20, ABCB1, Abcb1a, Abcb1b, Abcb4, ATP binding cassette subfamily B member 1, ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1), ATP-binding cassette sub-family B member 1, ATP-binding cassette sub-family B member 1A, ATP-binding cassette sub-family B member 1B, ATP-binding cassette transporter subfamily B, member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1), ATP-binding cassette, subfamily B (MDR/TAP), member 1A, ATP-binding cassette, sub-family B (MDR/TAP), member 1A, ATP-binding cassette, subfamily B (MDR/TAP), member 1B, ATP-binding cassette, sub-family B (MDR/TAP), member 1B, ATP-binding cassette, sub-family B (MDR/TAP), member 4, ATP-binding cassette, subfamily B, member 1A, ATP-binding cassette, subfamily B, member 1B, ATP-dependent translocase ABCB1, ATP-dependent translocase ABCB1, multidrug resistance protein 1, bovine p-glycoprotein, CD243, CLCS, colchicin sensitivity, doxorubicin resistance, ecotropic viral integration site 32, Evi32, GP170, mdr, Mdr1, MDR1A, Mdr1b, Mdr3, mdr-3, multi-drug resistance 3, multidrug resistance p-glycoprotein, multidrug resistance protein 1, Multidrug resistance protein 1A, multidrug resistance protein 1B, Multidrug resistance protein 2, multidrug resistance protein 3, multidrug resistance protein MDR1, multiple drug resistant 1a, P glycoprotein 1, P glycoprotein 3, P-glycoprotein, P-glycoprotein 1, P-glycoprotein 2, P-glycoprotein 3, P-glycoprotein/multidrug resistance 1, Pgp, p-gp, PGP1, PGP2, PGY1, Pgy-1, Pgy1-1, PGY2, Pgy3, Pgy-3, Phospholipid transporter ABCB1
Gene Symbols ABCB1, Abcb1a, Abcb1b
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 590-689 of human P Glycoprotein (NP_0009182)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170913, 18669, 18671, 24646, 5243
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.