Learn More
P-gp (CD243) Mouse, anti-Human, R718, Clone: 15D3, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752143
Description
The 15D3 monoclonal antibody specifically recognizes P-glycoprotein which is also known as CD243, ATP-binding cassette subfamily B member 1 (ABCB1) or Multidrug resistance protein 1 (MDR1). P-glycoprotein is a ∼170 kDa transmembrane glycoprotein protein that spans the membrane 12 times. P-glycoprotein acts as an ATP-dependent efflux pump for a large variety of drugs. It may be expressed at high levels by multidrug resistant (MDR) tumor cells. This efflux activity may lead to cellular resistance to the drugs used in chemotherapy. P-glycoprotein is present in many normal cell types including endothelial, epithelial, or secretory cells, and might protect them from naturally occurring xenobiotics.
Specifications
| P-gp (CD243) | |
| Monoclonal | |
| R718 | |
| 50 μg | |
| Primary | |
| Refrigerate Upon Arrival |
| Flow Cytometry | |
| 15D3 | |
| Mouse | |
| RUO | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.