Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Iodonitrobenzene, MP Biomedicals

Catalog No. ICN210574
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 636-98-6
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
InChI Key SCCCFNJTCDSLCY-UHFFFAOYSA-N
Synonym benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene
PubChem CID 12495
IUPAC Name 1-iodo-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])I

Specifications

Quantity 10 g
Formula Weight 249.01
Chemical Name or Material p-Iodonitrobenzene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.