Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Naphtholbenzein Indicator, 1% in Titration Solvent, Reagents

Catalog No. p-200021487
Click to view available options
Quantity:
120 mL
500 mL
1 L

p-Naphtholbenzein Indicator, 1% in Titration Solvent ASTM Methods D974, D3242, D3339, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material p-Naphtholbenzein Indicator, 1% in Titration Solvent ASTM Methods D974, D3242, D3339
Boiling Point 90°C
CAS 108-88-3, 67-63-0, 7732-18-5
Synonym alpha-Naphtholbenzein, para-Naphtholbenzein, 1-Naphtholbenzein
Grade ASTM
Packaging Glass Bottle
Specific Gravity 0.8245
Color Dark Red
Quantity 120 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.