Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Nitrocumene, 90%, MP Biomedicals

Catalog No. ICN222706
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 1817-47-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key JXMYUMNAEKRMIP-UHFFFAOYSA-N
Synonym 1-isopropyl-4-nitrobenzene, p-nitrocumene, 4-nitrocumene, 4-isopropylnitrobenzene, benzene, 1-1-methylethyl-4-nitro, cumene, p-nitro, p-isopropylnitrobenzene, p-nitroisopropylbenzene, 4-nitroisopropylbenzene, 2-p-nitrophenyl propane
PubChem CID 15749
IUPAC Name 1-nitro-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 10 g
Formula Weight 165.19
Chemical Name or Material p-Nitrocumene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.