Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Phenylenediamine, free base, 99.9%, MP Biomedicals™

Catalog No. ICN15183080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15183080 100 g
ICN15183050 50 g
ICN15183083 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15183080 Supplier MP Biomedicals, Inc Supplier No. 0215183080

Chemical Identifiers

CAS 106-50-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
InChI Key CBCKQZAAMUWICA-UHFFFAOYSA-N
Synonym p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
PubChem CID 7814
ChEBI CHEBI:51403
IUPAC Name benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N

Specifications

Assay Percent Range 0.999
Quantity 100 g
Formula Weight 108.1
Physical Form Crystalline
Chemical Name or Material p-Phenylenediamine, free base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.