Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Rex1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297481B
Description
P-Rex1 Polyclonal specifically detects P-Rex1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
P-Rex1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, ptdIns(34,5)-dependent Rac exchanger 1 | |
Produced in rabbits immunized with E. coli-derived Human P-Rex1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
57580 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction