Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Selectin/CD62P Antibody (001), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290703AF647
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Mouse samples. It is validated for ELISA.Specifications
P-Selectin/CD62P | |
Monoclonal | |
Alexa Fluor 647 | |
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse P-Selectin/CD62P (Q01102; Met1-Ala709). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
6403 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction