Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Selectin/CD62P Antibody (AK-4), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143392APCCY7
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry.Specifications
P-Selectin/CD62P | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
SELP | |
The immunogen for this antibody was CD62P. | |
0.1 mL | |
6403.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
AK-4 | |
Flow Cytometry | |
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
Mouse | |
Protein A or G purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction