Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Selectin/CD62P Antibody (AK-4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143392IR
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
P-Selectin/CD62P | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SELP | |
The immunogen for this antibody was CD62P. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
AK-4 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
Mouse | |
Protein A or G purified | |
RUO | |
6403 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction