Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Selectin/CD62P Antibody, Novus Biologicals™
SDP

Catalog No. p-7200076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018668 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018668 Supplier Novus Biologicals Supplier No. NBP1507640.025MG

Rabbit Polyclonal Antibody

P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen P-Selectin/CD62P
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL, Immunohistochemistry
Formulation PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Gene Accession No. Q01102
Gene Alias CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
Gene Symbols SELP
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 756 of mouse CD62P
Molecular Weight of Antigen 91 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Target Species Mouse, Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.