Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.12), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065391PCP
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human, Rat, Goat, Sheep samples. It is validated for Flow Cytometry.Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| PerCP | |
| P16109 | |
| SELP | |
| P-Selectin transfected 300.19 cells. | |
| 0.1 mL | |
| Primary | |
| NB100-65391 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. Clone Psel.KO.2.12 is reported to inhibit P-selectin-dependent adhesion between HL60 cells and P-selectin transfected COS cells. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Psel.KO.2.12 | |
| Flow Cytometry | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6403 | |
| Human, Rat, Goat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction