Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 14 publications
$206.50 - $499.50
Specifications
Antigen | P-Selectin/CD62P |
---|---|
Clone | Psel.KO.2.7 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB165392SS
![]() |
Novus Biologicals
NB10065392SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065392
![]() |
Novus Biologicals
NB10065392 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
P-Selectin/CD62P | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Bovine, Equine, Feline, Goat, Sheep | |
CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
SELP | |
IgG1 | |
Protein G purified | |
NB100-65392 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. |
Psel.KO.2.7 | |
Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
P16109 | |
6403 | |
P-selectin transfected 300.19 cells. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title