Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P-Selectin/CD62P Antibody (Psel.KO.2.12), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065391APCCY7
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human, Rat, Goat, Sheep samples. It is validated for Flow Cytometry.Specifications
| P-Selectin/CD62P | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| SELP | |
| P-Selectin transfected 300.19 cells. | |
| 0.1 mL | |
| 6403.0 | |
| Human, Rat, Goat, Sheep | |
| IgG1 |
| Flow Cytometry | |
| Psel.KO.2.12 | |
| Flow Cytometry | |
| CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62) | |
| Mouse | |
| Protein G purified | |
| Primary | |
| NB100-65391 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. Clone Psel.KO.2.12 is reported to inhibit P-selectin-dependent adhesion between HL60 cells and P-selectin transfected COS cells. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction