Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-selectin Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728639 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8639 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8639 Supplier Proteintech Group Inc Supplier No. 133041AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P-Selectin, also called CD62, GRMP, GMP140 is a 140-kD adhesion molecule, expressed at the surface of activated cells, that mediates the interaction of activated endothelial cells or platelets with leukocytes. The P-Selectin gene is mapped on 1q24.2. P-selectin plays an essential role in the initial recruitment of leukocytes (white blood cells) to the site of injury during inflammation. P-selectin is also very important in the recruitment and aggregation of platelets at areas of vascular injury. In quiescent platelet, P-selectin is located on the inner wall of alpha-granules. Sorting nexin-17 interacts with the cytosolic domain of P-selectin and suggested that SNX17 may function in the intracellular trafficking of P-selectin. P-selectin expression was found to be significantly and positively correlated with carotid artery stiffness and wall thickness, and the percentage of P-selectin-positive platelets was significantly higher in patients with carotid plaque.

Specifications

Antigen P-selectin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SELP
Gene Accession No. P16109
Gene Alias CD62, CD62P, GMP 140, GMP140, GMRP, Granule membrane protein 140, GRMP, LECAM3, P selectin, PADGEM, PSEL, P-selectin, SELP
Gene Symbols SELP
Host Species Rabbit
Immunogen P-selectin Fusion Protein Ag4124
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.