Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolualdehyde, 98%

Catalog No. p-7081416
Click to view available options
Quantity:
5 g
100 g
2500 g

CAS: 104-87-0 | C8H8O | 120.151 g/mol

p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-87-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00006954
InChI Key FXLOVSHXALFLKQ-UHFFFAOYSA-N
Synonym p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
PubChem CID 7725
ChEBI CHEBI:28617
IUPAC Name 4-methylbenzaldehyde
SMILES CC1=CC=C(C=C1)C=O

Specifications

Melting Point -6°C
Density 1.018
Boiling Point 204°C
Flash Point 80°C (176°F)
Refractive Index 1.546
Quantity 5 g
Beilstein 385772
Sensitivity Air Sensitive
Solubility Information It is slightly soluble in water.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material p-Tolualdehyde
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-246-9
RTECSNumber CU7034500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.